Learn More
Apexbio Technology LLC 1-Naphthaleneacetic acid 86-87-3 1g
SDP
Catalog No. 5000094643
Encompass_Preferred
1-Naphthaleneacetic acid (CAS 86-87-3) is a synthetic auxin belonging to the naphthalene organic compound family characterized by two fused benzene rings It functions as a plant growth regulator by mimicking the activity of natural auxins binding to auxin receptors and modulating gene expression involved in cell elongation division and differentiation In biomedical research 1-naphthaleneacetic acid is widely used to investigate auxin-mediated signaling pathways plant development and cellular responses to exogenous hormones offering a valuable tool for dissecting phytohormone functions and regulatory networks
Catalog No. 50-000-94643 Supplier Apexbio Technology LLC Supplier No. C71411000
Add to Cart
Add to Cart
1-Naphthaleneacetic acid (CAS 86-87-3) is a synthetic auxin belonging to the naphthalene organic compound family characterized by two fused benzene rings It functions as a plant growth regulator by mimicking the activity of natural auxins binding to auxin receptors and modulating gene expression involved in cell elongation division and differentiation In biomedical research 1-naphthaleneacetic acid is widely used to investigate auxin-mediated signaling pathways plant development and cellular responses to exogenous hormones offering a valuable tool for dissecting phytohormone functions and regulatory networks