Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthaleneacetic Acid, 97%, Spectrum™ Chemical
SDP

Catalog No. 18600064 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-064 500 g Poly Pail
18-600-062 25 g Poly Bottle
18-600-063 100 g Poly Bottle
18-600-065 1 kg Poly Pail
18-600-066 5 kg Poly Pail
18-600-067 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-600-064 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. NA107500GM
Only null left
Add to Cart
Add to Cart

NA107, 86-87-3, C12H10O2

Spectrum™ Chemical 1-Naphthaleneacetic Acid is an organic compound that is colorless and soluble in organic solvents. It is a synthetic plant hormone that is part of the auxin family and is found in most commercial horticultural plant rooting products and is used for rooting and propagation purposes.

Chemical Identifiers

CAS 86-87-3
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004046
InChI Key PRPINYUDVPFIRX-UHFFFAOYSA-N
IUPAC Name 2-(naphthalen-1-yl)acetic acid
SMILES OC(=O)CC1=C2C=CC=CC2=CC=C1

Specifications

CAS Max % 100%
Packaging Poly Pail
Quantity 500 g
Percent Purity 97%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.