Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Naphthaleneboronic acid | 13922-41-3 | 99.96% | C10H9BO2 | 100 G
SDP

Catalog No. 5000434564
Encompass_Preferred

1-Naphthaleneboronic acid is a biochemical reagent suitable for life science research. It serves as a biological material or organic compound, appearing as a white to off-white solid.

  • Biochemical reagent for life science research
  • Functions as a biological material or organic compound
  • Available as a solid
  • White to off-white appearance
  • Molecular weight: 171.99
  • Chemical formula: C10H9BO2

Catalog No. 50-004-34564 Supplier Medchemexpress LLC Supplier No. HY78494100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Naphthaleneboronic acid is a biochemical reagent suitable for life science research. It serves as a biological material or organic compound, appearing as a white to off-white solid.

  • Biochemical reagent for life science research
  • Functions as a biological material or organic compound
  • Available as a solid
  • White to off-white appearance
  • Molecular weight: 171.99
  • Chemical formula: C10H9BO2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.