Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-naphthaleneethanol | 773-99-9 | MFCD00004050 | 99.9% | 172.22 g/mol | C12H12O | 25 G
SDP

Catalog No. 5000304159
Encompass_Preferred

2-(Naphthalen-1-yl)ethanol, also known as 1-naphthaleneethanol, is an aromatic primary alcohol used as a building block and reagent in organic synthesis and life-science research. It is supplied at high purity for analytical use and small-scale synthesis, with documented storage and stability recommendations for both powder and solution forms.

  • High purity: 99.9% by HPLC (rounded from COA).
  • Chemical formula: C12H12O.
  • Molecular weight: 172.22 g/mol.
  • Intended use: for research use only and method development.
  • Storage and stability: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Common applications: organic synthesis, analytical reference, and building block chemistry.

Catalog No. 50-003-04159 Supplier Medchemexpress LLC Supplier No. 25GHYW016876
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Naphthalen-1-yl)ethanol, also known as 1-naphthaleneethanol, is an aromatic primary alcohol used as a building block and reagent in organic synthesis and life-science research. It is supplied at high purity for analytical use and small-scale synthesis, with documented storage and stability recommendations for both powder and solution forms.

  • High purity: 99.9% by HPLC (rounded from COA).
  • Chemical formula: C12H12O.
  • Molecular weight: 172.22 g/mol.
  • Intended use: for research use only and method development.
  • Storage and stability: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Common applications: organic synthesis, analytical reference, and building block chemistry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.