Learn More
Medchemexpress LLC 1-Naphthalenemethanamine, N-methyl-N-[(2E)-3-phenyl-2-propen-1-yl]-, hydrochloride (1:1) | 65473-14-5 | 99.7% | 100 MG
SDP
Catalog No. 5000407564
Encompass_Preferred
Naftifine hydrochloride is an antibiotic with antifungal activity against dermatophytes, aspergilli, Sporothrix schenckii, and yeasts of the genus Candida. It can be used for the research of superficial dermatomycoses inhibition.
  • Antifungal activity against dermatophytes
  • Antifungal activity against aspergilli
  • Antifungal activity against Sporothrix schenckii
  • Antifungal activity against yeasts of the genus Candida
  • Used for research of superficial dermatomycoses inhibition
Catalog No. 50-004-07564 Supplier Medchemexpress LLC Supplier No. HYB0518A100MG
Naftifine hydrochloride is an antibiotic with antifungal activity against dermatophytes, aspergilli, Sporothrix schenckii, and yeasts of the genus Candida. It can be used for the research of superficial dermatomycoses inhibition.
  • Antifungal activity against dermatophytes
  • Antifungal activity against aspergilli
  • Antifungal activity against Sporothrix schenckii
  • Antifungal activity against yeasts of the genus Candida
  • Used for research of superficial dermatomycoses inhibition