Learn More
Medchemexpress LLC 1-naphthalenesulfonic acid, 3,4-dihydro-3,4-dioxo-, potassium salt | 5908-27-0 | MFCD00001699 | 99.8% | 276.31 | C10H5KO5S | 1g
SDP
Catalog No. 5000270080
Encompass_Preferred
Potassium 1 2-naphthoquinone-4-sulphonate (Compound 16) is a 6-phosphogluconate dehydrogenase (6PGDH) inhibitor with an IC50 of 53 M Potassium 1 2-naphthoquinone-4-sulphonate can be used in the study of human African trypanosomiasis[1]
Catalog No. 50-002-70080 Supplier Medchemexpress LLC Supplier No. HYW7261371G
Potassium 1 2-naphthoquinone-4-sulphonate (Compound 16) is a 6-phosphogluconate dehydrogenase (6PGDH) inhibitor with an IC50 of 53 M Potassium 1 2-naphthoquinone-4-sulphonate can be used in the study of human African trypanosomiasis[1]