Learn More
Medchemexpress LLC 1-Naphthalenesulfonyl chloride, 5-[di(methyl-d3)amino]- | 1276379-68-0 | 98.0% | 1 MG
SDP
Catalog No. 5000410136
Encompass_Preferred
Dansyl chloride-d6 is the deuterium labeled form of Dansyl chloride, a reagent known for producing stable blue or blue-green fluorescent sulfonamide adducts. It reacts with aliphatic and aromatic amines containing primary amino groups and is widely utilized for modified amino acids, protein sequencing, and amino acid analysis. This compound serves as a valuable tracer and an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Applicable in NMR, GC-MS, or LC-MS
  • Deuterium-labeled compound
Catalog No. 50-004-10136 Supplier Medchemexpress LLC Supplier No. HYD0017S1MG
Dansyl chloride-d6 is the deuterium labeled form of Dansyl chloride, a reagent known for producing stable blue or blue-green fluorescent sulfonamide adducts. It reacts with aliphatic and aromatic amines containing primary amino groups and is widely utilized for modified amino acids, protein sequencing, and amino acid analysis. This compound serves as a valuable tracer and an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Applicable in NMR, GC-MS, or LC-MS
  • Deuterium-labeled compound