Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthalenethiol, 99%

Catalog No. ac415270050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC415270050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC415270050 Supplier Thermo Scientific Chemicals Supplier No. 415270050
Only null left
Add to Cart

CAS: 529-36-2 | C10H7S | 159.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-36-2
Molecular Formula C10H7S
Molecular Weight (g/mol) 159.23
MDL Number MFCD00039599
InChI Key SEXOVMIIVBKGGM-UHFFFAOYSA-M
Synonym 1-naphthalenethiol, 1-thionaphthol, naphthalenethiol, 1-naphthylthiol, unii-9a78cp495h, 1-mercaptonaphthalene, .alpha.-naphthyl mercaptan, naphthalene thiol, 1-napthalenethiol, naphthalen-1-thiol
PubChem CID 68259
IUPAC Name naphthalene-1-thiol
SMILES [S-]C1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.1500g/mL
Boiling Point 131°C (5.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6790 to 1.681
Quantity 5 g
Specific Gravity 1.15
Merck Index 15, 6463
Formula Weight 160.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Naphthalenethiol, 99%
Show More Show Less
EINECSNumber 208-462-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.