Learn More
Medchemexpress LLC 1-Naphthalenol, 4-[(7-chloro-4-quinolinyl)amino]-2-[[(1,1-dimethylethyl)amino]methyl]-5,6,7,8-tetrahydro-, phosphate (1:2) (salt) (9CI) | 173531-58-3 | MFCD00222014 | 99.59% | 605.94 | 50 MG
SDP
Catalog No. 5000384211
Encompass_Preferred
Naphthoquine phosphate is a potent and orally active antimalarial agent. It has thorough killing function for various schizonts of plasmodia, including resistance of P. falciparum to Chloroquine. For research use only. We do not sell to patients.
  • Potent and orally active antimalarial agent.
  • Thorough killing function for various schizonts of plasmodia, including resistance of P. falciparum to Chloroquine.
Catalog No. 50-003-84211 Supplier Medchemexpress LLC Supplier No. HY1703650MG
Naphthoquine phosphate is a potent and orally active antimalarial agent. It has thorough killing function for various schizonts of plasmodia, including resistance of P. falciparum to Chloroquine. For research use only. We do not sell to patients.
  • Potent and orally active antimalarial agent.
  • Thorough killing function for various schizonts of plasmodia, including resistance of P. falciparum to Chloroquine.