Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthoic hydrazide, 98+%

Catalog No. AAL0794214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0794214 25 g
AAL0794206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0794214 Supplier Thermo Scientific Chemicals Supplier No. L0794214
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 43038-45-5 | C11H10N2O | 186.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43038-45-5
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD00016804
InChI Key VMFUMDXVTKTZQY-UHFFFAOYSA-N
Synonym 1-naphthohydrazide, 1-naphthhydrazide, 1-naphthalenecarbohydrazide, 1-naphthalenecarboxylic acid, hydrazide, 1-naphthoic hydrazide, .alpha.-naphthoylhydrazine, naphthalene-1-carboxylic acid hydrazide, naphthalenecarbohydrazide, a-naphthoylhydrazine, 1-naphthohydrazide #
PubChem CID 72689
IUPAC Name naphthalene-1-carbohydrazide
SMILES C1=CC=C2C(=C1)C=CC=C2C(=O)NN

Specifications

Melting Point 166°C to 168°C
Quantity 25 g
Beilstein 2966714
Formula Weight 186.21
Percent Purity ≥98%
Chemical Name or Material 1-Naphthoic hydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 256-054-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.