Learn More
Medchemexpress LLC 1-naphthol-3,6,8-trisulfonic acid trisodium salt | 31894-34-5 | MFCD03427540 | 97.0% | 450.31 g/mol | C10H5Na3O10S3 | 10MG
SDP
Catalog No. 5000239414
Encompass_Preferred
ζ-Stat trisodium (1-naphthol-3,6,8-trisulfonic acid trisodium salt) is the trisodium salt research reagent that functions as a selective atypical protein kinase C-ζ (PKC-ζ) inhibitor with reported in vitro antitumor activity (IC50 ≈ 5 μM). It reduces proliferation and induces apoptosis in certain melanoma cell lines and is supplied as a solid for biochemical and cell-based assays.
  • Selective atypical PKC-ζ inhibitor (IC50 ≈ 5 μM).
  • Demonstrated antiproliferative and pro-apoptotic activity in melanoma cell lines.
  • Solid, light yellow appearance; purity 97.0%.
  • Water soluble: 12.5 mg/mL in H2O with ultrasonic/warming (heat to 60°C).
  • Store sealed, away from moisture; recommend 4°C; in solution keep at -80°C (6 months) or -20°C (1 month).
  • Available in small research pack sizes including 10 mg.
Catalog No. 50-002-39414 Supplier Medchemexpress LLC Supplier No. HY123979A10MG
Add to Cart
Add to Cart
ζ-Stat trisodium (1-naphthol-3,6,8-trisulfonic acid trisodium salt) is the trisodium salt research reagent that functions as a selective atypical protein kinase C-ζ (PKC-ζ) inhibitor with reported in vitro antitumor activity (IC50 ≈ 5 μM). It reduces proliferation and induces apoptosis in certain melanoma cell lines and is supplied as a solid for biochemical and cell-based assays.
  • Selective atypical PKC-ζ inhibitor (IC50 ≈ 5 μM).
  • Demonstrated antiproliferative and pro-apoptotic activity in melanoma cell lines.
  • Solid, light yellow appearance; purity 97.0%.
  • Water soluble: 12.5 mg/mL in H2O with ultrasonic/warming (heat to 60°C).
  • Store sealed, away from moisture; recommend 4°C; in solution keep at -80°C (6 months) or -20°C (1 month).
  • Available in small research pack sizes including 10 mg.