Learn More
Medchemexpress LLC HY-Y1309 5g Medchemexpress, 1-Naphthol CAS:90-15-3 Purity:>98%
SDP
Catalog No. 501934702
Encompass_Preferred
Medchemexpress, HY-Y1309 5g 1-Naphthol CAS:90-15-3 1-naphthol is an excited state proton transfer (ESPT) fluorescent molecular probe. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4702 Supplier Medchemexpress LLC Supplier No. HYY13095G
Medchemexpress, HY-Y1309 5g 1-Naphthol CAS:90-15-3 1-naphthol is an excited state proton transfer (ESPT) fluorescent molecular probe. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.