Learn More
Medchemexpress LLC 1-naphthol | 90-15-3 | MFCD00003930 | >99.0% | 50g
SDP
Catalog No. 5000266676
Encompass_Preferred
1-naphthol (Furro ER) is an excited state proton transfer (ESPT) fluorescent molecular probe 1-naphthol can sense the hydrophobic domains in micellar aggregation and accurately sense the sol-gel transition 1-naphthol can be used to study the thermally induced microenvironmental changes and hydration levels of Pluronic F127 (PF127) in different microenvironments in aqueous media[1]
Catalog No. 50-002-66676 Supplier Medchemexpress LLC Supplier No. HYY130950G
1-naphthol (Furro ER) is an excited state proton transfer (ESPT) fluorescent molecular probe 1-naphthol can sense the hydrophobic domains in micellar aggregation and accurately sense the sol-gel transition 1-naphthol can be used to study the thermally induced microenvironmental changes and hydration levels of Pluronic F127 (PF127) in different microenvironments in aqueous media[1]