Learn More
Cayman Chemical 1'-Naphthoyl Indole, Purity: ≥98%, CAS: 109555-87-5, 25 mg
SDP
Catalog No. 502952587
Encompass_Preferred
An analytical reference standard categorized as a precursor in the synthesis of synthetic cannabinoids; intended for research and forensic applications
Catalog No. 50-295-2587 Supplier Cayman Chemical Supplier No. 1168725 MG
An analytical reference standard categorized as a precursor in the synthesis of synthetic cannabinoids; intended for research and forensic applications