Learn More
Medchemexpress LLC 1-Naphthyl 3 5-dinit | 93261-39-3 | 10MG
SDP
Catalog No. 5000236540
Encompass_Preferred
1-Naphthyl 3 5-dinitrobenzoate is a potent 5-lipoxygenase inhibitor with IC50 values of 1 04 M and 3 6 M for 5-LOX and mPGES-1 respectively 1-Naphthyl 3 5-dinitrobenzoate has strong inhibition activity in the human whole blood (HWB) assay with an IC50 value of 8 6 M 1-Naphthyl 3 5-dinitrobenzoate can be used in research of inflammation[1]
Catalog No. 50-002-36540 Supplier Medchemexpress LLC Supplier No. HY12597510MG
1-Naphthyl 3 5-dinitrobenzoate is a potent 5-lipoxygenase inhibitor with IC50 values of 1 04 M and 3 6 M for 5-LOX and mPGES-1 respectively 1-Naphthyl 3 5-dinitrobenzoate has strong inhibition activity in the human whole blood (HWB) assay with an IC50 value of 8 6 M 1-Naphthyl 3 5-dinitrobenzoate can be used in research of inflammation[1]