Learn More
Medchemexpress LLC 1-naphthyl 3,5-dinitrobenzoate | 93261-39-3 | MFCD00376091 | 99.5% | 338.27 g/mol | C17H10N2O6 | 100MG
SDP
Catalog No. 5000238067
Encompass_Preferred
1-Naphthyl 3,5-dinitrobenzoate is a small-molecule inhibitor reported to target 5-lipoxygenase (5-LOX). It is supplied as a crystalline off-white to light-yellow solid with molecular formula C17H10N2O6 and molecular weight 338.27 g/mol. Typical supplied purity is approximately 99.5%, and the compound is packaged in milligram quantities for biochemical research use.
  • Potent inhibitor of 5-lipoxygenase (biochemical activity).
  • High purity suitable for enzymatic and cellular assays.
  • Crystalline off-white to light-yellow solid for easy handling and weighing.
  • Molecular formula C17H10N2O6 and molecular weight 338.27 g/mol.
  • Available in milligram-scale quantities for laboratory research.
Catalog No. 50-002-38067 Supplier Medchemexpress LLC Supplier No. HY125975100MG
1-Naphthyl 3,5-dinitrobenzoate is a small-molecule inhibitor reported to target 5-lipoxygenase (5-LOX). It is supplied as a crystalline off-white to light-yellow solid with molecular formula C17H10N2O6 and molecular weight 338.27 g/mol. Typical supplied purity is approximately 99.5%, and the compound is packaged in milligram quantities for biochemical research use.
  • Potent inhibitor of 5-lipoxygenase (biochemical activity).
  • High purity suitable for enzymatic and cellular assays.
  • Crystalline off-white to light-yellow solid for easy handling and weighing.
  • Molecular formula C17H10N2O6 and molecular weight 338.27 g/mol.
  • Available in milligram-scale quantities for laboratory research.