Learn More
Medchemexpress LLC 1-naphthyl 3,5-dinitrobenzoate | 93261-39-3 | MFCD00376091 | >=98.0% | 338.27 g/mol | C17H10N2O6 | 1MG
SDP
Catalog No. 5000238335
Encompass_Preferred
1-Naphthyl 3,5-dinitrobenzoate is a small-molecule biochemical reagent used as a dual inhibitor of 5-lipoxygenase (5-LOX) and microsomal prostaglandin E synthase-1 (mPGES-1). Supplied as a crystalline solid, it is used in inflammation and lipid mediator research to probe enzyme activity and signaling pathways. Molecular formula C17H10N2O6; molecular weight 338.27 g/mol; reported IC50 values ≈1 μM (5-LOX) and ≈3.6 μM (mPGES-1).
  • Potent dual inhibitor for use in target validation and pathway studies.
  • High-purity crystalline solid suitable for biochemical assays.
  • Well characterized molecular formula and molecular weight for analytical work.
  • Stable when stored at low temperature for extended shelf life.
  • Available in small research quantities for lab-scale experiments.
Catalog No. 50-002-38335 Supplier Medchemexpress LLC Supplier No. HY1259751MG
1-Naphthyl 3,5-dinitrobenzoate is a small-molecule biochemical reagent used as a dual inhibitor of 5-lipoxygenase (5-LOX) and microsomal prostaglandin E synthase-1 (mPGES-1). Supplied as a crystalline solid, it is used in inflammation and lipid mediator research to probe enzyme activity and signaling pathways. Molecular formula C17H10N2O6; molecular weight 338.27 g/mol; reported IC50 values ≈1 μM (5-LOX) and ≈3.6 μM (mPGES-1).
  • Potent dual inhibitor for use in target validation and pathway studies.
  • High-purity crystalline solid suitable for biochemical assays.
  • Well characterized molecular formula and molecular weight for analytical work.
  • Stable when stored at low temperature for extended shelf life.
  • Available in small research quantities for lab-scale experiments.