Learn More
Medchemexpress LLC 1-naphthyl 3,5-dinitrobenzoate | 93261-39-3 | MFCD00376091 | 99.5% | 338.27 g·mol⁻¹ | C17H10N2O6 | 5MG
SDP
Catalog No. 5000237817
Encompass_Preferred
1-Naphthyl 3,5-dinitrobenzoate (CAS 93261-39-3) is a small-molecule research compound reported as a 5-lipoxygenase (5-LOX) inhibitor used in inflammation research. It is an off-white to light-yellow solid with formula C17H10N2O6 and molecular weight 338.27 g·mol⁻1. Typical purity is listed at 99.49% and the compound is supplied in milligram quantities for in vitro studies.
  • Potent 5-LOX inhibitor with reported IC50 of 1.04 μM.
  • Also reported to inhibit mPGES-1 with reported IC50 of 3.6 μM.
  • Off-white to light-yellow solid suitable for in vitro biochemical assays.
  • High purity for research applications (≈99.5%).
  • Supplied in milligram-scale quantities for laboratory studies.
Catalog No. 50-002-37817 Supplier Medchemexpress LLC Supplier No. HY1259755MG
1-Naphthyl 3,5-dinitrobenzoate (CAS 93261-39-3) is a small-molecule research compound reported as a 5-lipoxygenase (5-LOX) inhibitor used in inflammation research. It is an off-white to light-yellow solid with formula C17H10N2O6 and molecular weight 338.27 g·mol⁻1. Typical purity is listed at 99.49% and the compound is supplied in milligram quantities for in vitro studies.
  • Potent 5-LOX inhibitor with reported IC50 of 1.04 μM.
  • Also reported to inhibit mPGES-1 with reported IC50 of 3.6 μM.
  • Off-white to light-yellow solid suitable for in vitro biochemical assays.
  • High purity for research applications (≈99.5%).
  • Supplied in milligram-scale quantities for laboratory studies.