Learn More
Selleck Chemical LLC 1-Naphthyl acetate S4804-25mg
SDP
Catalog No. 503108184
Encompass_Preferred
1-Naphthyl acetate is usually used in a rapid staining method for identification of macrophages 1-Naphthyl acetate is a potent chromogenic substrate for the detection of erythrocyte acetylcholinesterase (AChE) activity
Catalog No. 50-310-8184 Supplier Selleck Chemical LLC Supplier No. S480425MG
1-Naphthyl acetate is usually used in a rapid staining method for identification of macrophages 1-Naphthyl acetate is a potent chromogenic substrate for the detection of erythrocyte acetylcholinesterase (AChE) activity