Learn More
Medchemexpress LLC 1-Naphthyl acetate | 830-81-9 | MFCD00003922 | 99.7% | 186.21 g/mol | C12H10O2 | 1 G
SDP
Catalog No. 5000302177
Encompass_Preferred
1-Naphthyl acetate is a chromogenic acetylcholinesterase (AChE) substrate used to detect AChE and esterase activity in biological samples. It is provided in solid and solution formats (for example, 10 mM in DMSO) for enzymatic assays and analytical applications; molecular formula C12H10O2, molecular weight 186.21, CAS 830-81-9. For research use only.
  • Chromogenic substrate for acetylcholinesterase and esterase assays.
  • Available as powder and ready-to-use DMSO solution.
  • High purity for consistent, reliable assay results.
  • Suitable for enzymatic activity detection and staining applications.
  • Supplied in multiple pack sizes to support varied experimental scales.
Catalog No. 50-003-02177 Supplier Medchemexpress LLC Supplier No. HYW0161881G
1-Naphthyl acetate is a chromogenic acetylcholinesterase (AChE) substrate used to detect AChE and esterase activity in biological samples. It is provided in solid and solution formats (for example, 10 mM in DMSO) for enzymatic assays and analytical applications; molecular formula C12H10O2, molecular weight 186.21, CAS 830-81-9. For research use only.
  • Chromogenic substrate for acetylcholinesterase and esterase assays.
  • Available as powder and ready-to-use DMSO solution.
  • High purity for consistent, reliable assay results.
  • Suitable for enzymatic activity detection and staining applications.
  • Supplied in multiple pack sizes to support varied experimental scales.