Learn More
Medchemexpress LLC 1-naphthyl acetate | 830-81-9 | MFCD00003922 | 99.7% | 186.21 g/mol | C12H10O2 | 1 ML
SDP
Catalog No. 5000302668
Encompass_Preferred
1-Naphthyl acetate is a chromogenic substrate for detection of acetylcholinesterase activity. Supplied as a ready-to-use 10 mM solution in DMSO, it is intended for biochemical assays requiring sensitive colorimetric readouts.
  • Ready-to-use 10 mM solution in DMSO.
  • Chromogenic substrate for acetylcholinesterase activity assays.
  • High purity (99.71%) for consistent assay performance.
  • Small 1 mL pack size suitable for limited-use experiments.
  • Molecular formula C12H10O2 and molecular weight 186.21 g/mol for reference.
Catalog No. 50-003-02668 Supplier Medchemexpress LLC Supplier No. HYW01618810MM/1ML
1-Naphthyl acetate is a chromogenic substrate for detection of acetylcholinesterase activity. Supplied as a ready-to-use 10 mM solution in DMSO, it is intended for biochemical assays requiring sensitive colorimetric readouts.
  • Ready-to-use 10 mM solution in DMSO.
  • Chromogenic substrate for acetylcholinesterase activity assays.
  • High purity (99.71%) for consistent assay performance.
  • Small 1 mL pack size suitable for limited-use experiments.
  • Molecular formula C12H10O2 and molecular weight 186.21 g/mol for reference.