Learn More
Medchemexpress LLC 1-naphthyl acetate | 830-81-9 | MFCD00003922 | 99.7% | 186.21 g/mol | C12H10O2 | 25 G
SDP
Catalog No. 5000304152
Encompass_Preferred
1-Naphthyl acetate is a small-molecule chromogenic reagent used as a substrate to measure acetylcholinesterase (AChE) activity in biochemical assays. Upon enzymatic hydrolysis it yields a colored product, enabling quantitative and qualitative detection in clinical, research, and environmental testing for organophosphorus pesticide exposure.
  • Chromogenic substrate for acetylcholinesterase assays.
  • Suitable for erythrocyte and tissue AChE activity measurements.
  • Enables detection of organophosphorus pesticide exposure.
  • High purity (99.7%), supporting reproducible assay results.
  • Available in multiple laboratory pack sizes for assay flexibility.
Catalog No. 50-003-04152 Supplier Medchemexpress LLC Supplier No. 25GHYW016188
1-Naphthyl acetate is a small-molecule chromogenic reagent used as a substrate to measure acetylcholinesterase (AChE) activity in biochemical assays. Upon enzymatic hydrolysis it yields a colored product, enabling quantitative and qualitative detection in clinical, research, and environmental testing for organophosphorus pesticide exposure.
  • Chromogenic substrate for acetylcholinesterase assays.
  • Suitable for erythrocyte and tissue AChE activity measurements.
  • Enables detection of organophosphorus pesticide exposure.
  • High purity (99.7%), supporting reproducible assay results.
  • Available in multiple laboratory pack sizes for assay flexibility.