Learn More
Medchemexpress LLC 1-naphthyl acetate | 830-81-9 | MFCD00003922 | 99.7% | 186.21 g/mol | C12H10O2 | 50 G
SDP
Catalog No. 5000303247
Encompass_Preferred
1-Naphthyl acetate is a chromogenic substrate for measuring acetylcholinesterase (AChE) activity. It yields a colored product upon enzymatic hydrolysis, enabling quantitative assays for enzymology and screening for organophosphorus pesticide inhibition. The compound is supplied as a high-purity solid with recommended storage conditions to preserve stability.
  • Chromogenic substrate for acetylcholinesterase assays.
  • Enables detection of organophosphorus pesticide inhibition.
  • High purity suitable for analytical and research use.
  • Available in multiple pack sizes to suit experimental scale.
  • Powder form with defined storage recommendations for stability.
  • Molecular data provided to support formulation and calculations.
Catalog No. 50-003-03247 Supplier Medchemexpress LLC Supplier No. HYW01618850G
1-Naphthyl acetate is a chromogenic substrate for measuring acetylcholinesterase (AChE) activity. It yields a colored product upon enzymatic hydrolysis, enabling quantitative assays for enzymology and screening for organophosphorus pesticide inhibition. The compound is supplied as a high-purity solid with recommended storage conditions to preserve stability.
  • Chromogenic substrate for acetylcholinesterase assays.
  • Enables detection of organophosphorus pesticide inhibition.
  • High purity suitable for analytical and research use.
  • Available in multiple pack sizes to suit experimental scale.
  • Powder form with defined storage recommendations for stability.
  • Molecular data provided to support formulation and calculations.