Learn More
Medchemexpress LLC 1-Naphthyl-d7 N-methylcarbamate | 362049-56-7 | 99% | 5 MG
SDP
Catalog No. 5000411978
Encompass_Preferred
Carbaryl-d7 is a deuterium-labeled version of Carbaryl, an acetylcholinesterase inhibitor. It functions by preventing the breakdown of acetylcholine in the synaptic cleft, leading to acetylcholine accumulation and neurotoxicity. This compound is primarily used as an insecticide.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis using NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes for pharmacokinetic and metabolic studies.
Catalog No. 50-004-11978 Supplier Medchemexpress LLC Supplier No. HYB1315S15MG
Carbaryl-d7 is a deuterium-labeled version of Carbaryl, an acetylcholinesterase inhibitor. It functions by preventing the breakdown of acetylcholine in the synaptic cleft, leading to acetylcholine accumulation and neurotoxicity. This compound is primarily used as an insecticide.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis using NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes for pharmacokinetic and metabolic studies.