Learn More
Research Products International Corp 1-Naphthyl Phosphate Monosodium Salt Monohydrate, 25 Grams
SDP
Catalog No. 50213560
Encompass_Preferred
Substrate for determaination of phosphatase. Protect from moisture.
Catalog No. 50-213-560 Supplier Research Products International Corp Supplier No. N4204025.0
Substrate for determaination of phosphatase. Protect from moisture.