Learn More
Research Products International Corp 1-Naphthyl Phosphate Monosodium Salt Monohydrate, 5 Grams
SDP
Catalog No. 50213561
Encompass_Preferred
Substrate for determaination of phosphatase. Protect from moisture.
Catalog No. 50-213-561 Supplier Research Products International Corp Supplier No. N420405.0
Substrate for determaination of phosphatase. Protect from moisture.