Learn More
Selleck Chemical LLC 1-Naphthyl phosphate potassium salt S5441-25mg
SDP
Catalog No. 503109882
Encompass_Preferred
1-Naphthyl phosphate potassium salt ( -Naphthyl acid phosphate monopotassium salt) is a non-specific phosphatase inhibitor which acts on acid alkaline and protein phosphatases
Catalog No. 50-310-9882 Supplier Selleck Chemical LLC Supplier No. S544125MG
1-Naphthyl phosphate potassium salt ( -Naphthyl acid phosphate monopotassium salt) is a non-specific phosphatase inhibitor which acts on acid alkaline and protein phosphatases