Learn More
Alkali Scientific 1 Naphthyl Phosphate Monosodium Salt Monohydrate, 1 G
SDP
Catalog No. 5000661431
Encompass_Preferred
Naphthyl Phosphate Monosodium Salt Monohydrate is a highly purified enzyme substrate used in a variety of biochemical and enzymatic assays. It is commonly utilized for measuring enzyme activity, especially for phosphatase assays. The compound contains the monosodium salt form of naphthyl phosphate, which is hydrolyzed by enzymes, releasing a colored product that can be detected through spectrophotometry. This reagent is essential for research in biochemistry, molecular biology, and enzyme kinetics. It is supplied in a 1g package and is ideal for use in laboratory applications requiring precise enzyme-substrate reactions for analytical studies.
Catalog No. 50-006-61431 Supplier Alkali Scientific Supplier No. NAP002CP
Naphthyl Phosphate Monosodium Salt Monohydrate is a highly purified enzyme substrate used in a variety of biochemical and enzymatic assays. It is commonly utilized for measuring enzyme activity, especially for phosphatase assays. The compound contains the monosodium salt form of naphthyl phosphate, which is hydrolyzed by enzymes, releasing a colored product that can be detected through spectrophotometry. This reagent is essential for research in biochemistry, molecular biology, and enzyme kinetics. It is supplied in a 1g package and is ideal for use in laboratory applications requiring precise enzyme-substrate reactions for analytical studies.