Learn More
Alkali Scientific 1 Naphthyl Phosphate Monosodium Salt Monohydrate, 25 Grams
SDP
Catalog No. 5000661370
Encompass_Preferred
Naphthyl Phosphate Monosodium Salt Monohydrate is a reagent commonly used as an enzyme substrate in biochemical research. It is particularly useful in assays that involve enzyme activity detection and metabolic studies. Naphthyl phosphate is hydrolyzed by various enzymes, releasing a colored product that can be quantified spectrophotometrically. This reagent is used in studies focusing on phosphatase activity, including alkaline phosphatase assays and other enzyme-catalyzed reactions. The 25g pack size ensures ample supply for routine laboratory use, making it an essential component for researchers investigating enzymatic activity, signal transduction pathways, and cellular processes in both academic and commercial labs.
Catalog No. 50-006-61370 Supplier Alkali Scientific Supplier No. NAP001CP
Naphthyl Phosphate Monosodium Salt Monohydrate is a reagent commonly used as an enzyme substrate in biochemical research. It is particularly useful in assays that involve enzyme activity detection and metabolic studies. Naphthyl phosphate is hydrolyzed by various enzymes, releasing a colored product that can be quantified spectrophotometrically. This reagent is used in studies focusing on phosphatase activity, including alkaline phosphatase assays and other enzyme-catalyzed reactions. The 25g pack size ensures ample supply for routine laboratory use, making it an essential component for researchers investigating enzymatic activity, signal transduction pathways, and cellular processes in both academic and commercial labs.