Learn More
Alkali Scientific 1 Naphthyl Phosphate Monosodium Salt Monohydrate, 5 G
SDP
Supplier:  Alkali Scientific NAP003CP
Encompass_Preferred
Naphthyl Phosphate Monosodium Salt Monohydrate (5g) is a high-purity enzyme substrate used for detecting phosphatase activity in various biochemical applications. When the substrate is hydrolyzed by phosphatase enzymes, it releases a colored product that can be measured spectrophotometrically. This compound is commonly used in enzyme assays to study enzyme kinetics and activity. The 5g package is suitable for large-scale experiments and multiple applications in research. It is ideal for use in biochemical, molecular biology, and clinical research labs that require precise substrate reactions for enzyme analysis.
Catalog No. 50-006-61326