Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthyl Phosphate Monosodium Salt Monohydrate, MP Biomedicals™

Catalog No. 0219941001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-199-41001 1 g
02-199-41005 5 g
02-199-41010 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-199-41001 Supplier MP Biomedicals, Inc Supplier No. 0219941001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inhibits acid, alkaline and protein phosphatases. 1-Naphthyl Phosphate Monosodium Salt Monohydrate, MP Biomedicals is a non-specific phosphatase inhibitor. It is the preferred substrate for bioanalytical determinations of acid phosphatases, such as prosta

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81012-89-7
Molecular Formula C10H7O4P
Molecular Weight (g/mol) 222.14
MDL Number MFCD00150615
InChI Key YNXICDMQCQPQEW-UHFFFAOYSA-L
Synonym 1-naphthyl phosphate monosodium salt monohydrate, sodium naphthalen-1-yl hydrogenphosphate hydrate, alpha-naphthyl acid phosphate monosodium, sodium hydrate naphthalen-1-yl hydrogen phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h8po4.na.h2o, sodium 1-naphthyl phosphate monohydrate, 1-naphthyl phosphate, monosodium salt monohydrate, naphthalen-1-yloxy sodiooxy phosphinic acid hydrate, 1-naphthyl phosphate monosodium salt monohydrate hplc
PubChem CID 45055387
SMILES [O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

Specifications

Physical Form Powder
Quantity 1 g
Solubility Information Soluble in water (50mg/mL)
IUPAC Name naphthalen-1-yl phosphate
Formula Weight 264.87g/mol
Chemical Name or Material 1-Naphthyl Phosphate Monosodium Salt Monohydrate
Recommended Storage Store at 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.