Learn More
TARGETMOL CHEMICALS INC 1-NAPHTHYL PP1 10MG
SDP
Catalog No. 502576855
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1-Naphthyl PP1 (1-NA-PP 1) is a selective src inhibitor(v-Src and c-Fyn c-Abl CDK2 and CAMK II with IC50s of 1.0 0.6 0.6 18 and 22 uM respectively)purity: 99%
Catalog No. 50-257-6855 Supplier TARGETMOL CHEMICALS INC Supplier No. T393510MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1-Naphthyl PP1 (1-NA-PP 1) is a selective src inhibitor(v-Src and c-Fyn c-Abl CDK2 and CAMK II with IC50s of 1.0 0.6 0.6 18 and 22 uM respectively)purity: 99%