Learn More
Medchemexpress LLC HY-13941B 5mg Medchemexpress, 1-Naphthyl PP1 (hydrochloride) CAS:956025-47-1 Purity:>98%
SDP
Catalog No. 502081534
Encompass_Preferred
Medchemexpress, HY-13941B 5mg 1-Naphthyl PP1 (hydrochloride) CAS:956025-47-1 1-Naphthyl PP1 hydrochloride (1-NA-PP 1 hydrochloride) is a selective inhibitor of src family kinases. 1-Naphthyl PP1 hydrochloride inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1534 Supplier Medchemexpress LLC Supplier No. HY13941B5MG
Medchemexpress, HY-13941B 5mg 1-Naphthyl PP1 (hydrochloride) CAS:956025-47-1 1-Naphthyl PP1 hydrochloride (1-NA-PP 1 hydrochloride) is a selective inhibitor of src family kinases. 1-Naphthyl PP1 hydrochloride inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.