Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthyl Propionate, TCI America™
SDP

Catalog No. P13845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13845G Supplier TCI America Supplier No. P13845G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3121-71-9
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00010496
InChI Key FRXPDEZCWCPLIH-UHFFFAOYSA-N
Synonym 1-naphthyl propionate, naphthalen-1-yl propionate, 1-naphthylpropionate, propionic acid 1-naphthyl ester, 1-naphthalenol, propanoate, naphthyl propanoate, naphthyl propionate, 1-naphthyl propanoate, 1-naphthalenol,1-propanoate, acmc-1bn98
PubChem CID 76572
IUPAC Name naphthalen-1-yl propanoate
SMILES CCC(=O)OC1=CC=CC2=CC=CC=C21

Specifications

Boiling Point 135°C
Quantity 5 g
Formula Weight 200.24
Chemical Name or Material 1-Naphthyl Propionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.