Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Naphthylacetamide, 98%

Click to view available options
Quantity:
25g
100g
500g
86-86-2
C12H11NO
185.226
MFCD00004047
XFNJVKMNNVCYEK-UHFFFAOYSA-N
1-naphthaleneacetamide,1-naphthylacetamide,2-1-naphthyl acetamide,frufix,rootone,naphthaleneacetamide,amid-thin,dirigol n,amid-thin w,naam
6861
CHEBI:81810
2-naphthalen-1-ylacetamide
C1=CC=C2C(=C1)C=CC=C2CC(=O)N
86-86-2
C12H11NO
185.226
MFCD00004047
XFNJVKMNNVCYEK-UHFFFAOYSA-N
1-naphthaleneacetamide,1-naphthylacetamide,2-1-naphthyl acetamide,frufix,rootone,naphthaleneacetamide,amid-thin,dirigol n,amid-thin w,naam
6861
CHEBI:81810
2-naphthalen-1-ylacetamide
C1=CC=C2C(=C1)C=CC=C2CC(=O)N
Melting Point 176°C to 178°C
Quantity 500g
Formula Weight 185.23
Percent Purity 98%
Chemical Name or Material 1-Naphthylacetamide

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA