Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Naphthylmethylamine

Catalog No. 12703525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-703-525G 25 g
12-703-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-703-525G Supplier Sigma Aldrich Supplier No. 12703525G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-naphthylmethylamine increases the induced circular dichroism (icd) magnitude exhibited by poly[(4-carboxyphenyl)acetylene]
  • It forms carbamate by reacting with monomethoxypoly(ethylene glycol) succinimido carbonate (mpeg-sc)

Specifications

CAS 118-31-0
Density 1.073 g/mL (at 25°C (literature))
Boiling Point 290°C to 293°C (lit.)
Molecular Formula C11H11N
Refractive Index n20/D 1.643 (literature)
Linear Formula C10H7CH2NH2
MDL Number MFCD00004048
Quantity 25 g
Synonym 1-(Aminomethyl)naphthalene; 1-Naphthalenemethylamine
Molecular Weight (g/mol) 157.21
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.