Learn More
Medchemexpress LLC HY-W011641 100mg Medchemexpress, (±)-Naringenin CAS:67604-48-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW011641100MG
Encompass_Preferred
Medchemexpress, HY-W011641 100mg (±)-Naringenin CAS:67604-48-2 (±)-Naringenin is a naturally-occurring flavonoid. (±)-Naringenin displays vasorelaxant effect on endothelium-denuded vessels via the activation of BKCa channels in myocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0685