Learn More
AdipoGen (±)-Naringenin (5 g)
SDP
Supplier:  AdipoGen CDXN0206G005
Encompass_Preferred
Chemical. CAS: 67604-48-2. Formula: C15H12O5. MW: 272.25. Naringenin is a citrus-derived flavonoid that inhibits CYP3A4 activity in human liver microsomes. It has a broad panel of properties, including antibacterial, antiviral, antifungal, anti-inflammatory, antioxidant, anticancer, antidiabetic, cardioprotective and neuroprotective activities. Naringenin activity is primarily attributed to its anti-inflammatory (via inhibiting recruitment of cytokines and inflammatory transcription factors) and antioxidant (via scavenging of free radicals, bolstering of endogenous antioxidant defense system and metal ion chelation) effects. Naringenin exerts its anti-diabetic effects by inhibition of gluconeogenesis through upregulations of AMPK, leading to its lipid lowering and insulin-like properties.
Catalog No. 50-205-4557