Learn More
Medchemexpress LLC 1-Nitropyrene 10g
SDP
Catalog No. 5000205777
Encompass_Preferred
1-Nitropyrene is a DNA probe whose metabolites can form nucleic acid adducts with DNA[1]
Catalog No. 50-002-05777 Supplier Medchemexpress LLC Supplier No. HYW09475510G
1-Nitropyrene is a DNA probe whose metabolites can form nucleic acid adducts with DNA[1]