Learn More
Medchemexpress LLC 1-nitrosopyrrolidine | 930-55-2 | MFCD00003166 | 100.0% | 100.12 g/mol | C4H8N2O | 50 G
SDP
Catalog No. 5000283322
Encompass_Preferred
1-Nitrosopyrrolidine is a small nitrosamine research reagent used to investigate DNA and protein alkylation and adduct formation in biochemical and toxicological studies. It is employed as a tool compound for mutagenesis, analytical method development, and mechanistic investigations of alkylation pathways.
  • Used to study DNA and protein adduct formation in vitro and in vivo
  • Small, well-characterized nitrosamine reactive with nucleic acids and proteins
  • High purity (99.98%) suitable for analytical and experimental use
  • Available in multiple pack sizes, including 50 G
  • Molecular identity: C4H8N2O, molecular weight 100.12 g/mol
Catalog No. 50-002-83322 Supplier Medchemexpress LLC Supplier No. HYW14107750G
Add to Cart
Add to Cart
1-Nitrosopyrrolidine is a small nitrosamine research reagent used to investigate DNA and protein alkylation and adduct formation in biochemical and toxicological studies. It is employed as a tool compound for mutagenesis, analytical method development, and mechanistic investigations of alkylation pathways.
  • Used to study DNA and protein adduct formation in vitro and in vivo
  • Small, well-characterized nitrosamine reactive with nucleic acids and proteins
  • High purity (99.98%) suitable for analytical and experimental use
  • Available in multiple pack sizes, including 50 G
  • Molecular identity: C4H8N2O, molecular weight 100.12 g/mol