Learn More
Selleck Chemical LLC 1-NM-PP1-5mg
SDP
Catalog No. 502389698
Encompass_Preferred
1-NM-PP1 (PP1 Analog II, 1NM-PP1, analogue 9) is a potent inhibitor of Src family kinases with IC50 of 4.3 nM and 3.2 nM for v-Src-as1 and c-Fyn-as1, respectively. 1-NM-PP1 also inhibits CDK2-as1, CAMKII-as1 and c-Abl-as2 with IC50 of 5.0 nM, 8.0 nM and 120 nM, respectively.
Catalog No. 50-238-9698 Supplier Selleck Chemical LLC Supplier No. S63835MG
1-NM-PP1 (PP1 Analog II, 1NM-PP1, analogue 9) is a potent inhibitor of Src family kinases with IC50 of 4.3 nM and 3.2 nM for v-Src-as1 and c-Fyn-as1, respectively. 1-NM-PP1 also inhibits CDK2-as1, CAMKII-as1 and c-Abl-as2 with IC50 of 5.0 nM, 8.0 nM and 120 nM, respectively.