Learn More
Apexbio Technology LLC 1-NM-PP1, 221244-14-0, MFCD17010191, 5 mg
SDP
Supplier:  Apexbio Technology LLC B12995
Encompass_Preferred
MF: C20H21N5, MW: 331.41, Solubility: 16.55mg/mL in DMSO, 9.22 mg/mL in EtOH with ultrasonic and warming,insoluble in H2O. 1-NM-PP1 is a C3-modified analog of cell-permeable PP1, acts as a potent and selective inhibitor of protein kinase families.
Catalog No. 50-115-1253