Learn More
Ace Glass, Inc. PTFE insert, 1" NPT, #25 compression fitting24 to 25mm OD tubing
SDP
Catalog No. 50977910
Encompass
Compression fitting PTFE inserts for 1NPT to #25 Ace-Thred. Fits 24 to 25mm O.D. tubing. For use with ACE PTFE reaction vessel heads.
Catalog No. 50-977-910 Supplier Ace Glass, Inc. Supplier No. 1287125
Compression fitting PTFE inserts for 1NPT to #25 Ace-Thred. Fits 24 to 25mm O.D. tubing. For use with ACE PTFE reaction vessel heads.