Learn More
Medchemexpress LLC 1-Nonadecanol | 1454-84-8 | 284.52 | 1 G
SDP
Catalog No. 5000448906
Encompass_Preferred
1-Nonadecanol is a component of the supercritical carbon dioxide (SC-CO2) essential oil of Heracleum thomsonii. It is also an important aroma compound found in Neotinea ustulata.
  • Component of SC-CO2 essential oil of Heracleum thomsonii.
  • Important aroma compound found in Neotinea ustulata.
Catalog No. 50-004-48906 Supplier Medchemexpress LLC Supplier No. HYW0042971G
1-Nonadecanol is a component of the supercritical carbon dioxide (SC-CO2) essential oil of Heracleum thomsonii. It is also an important aroma compound found in Neotinea ustulata.
  • Component of SC-CO2 essential oil of Heracleum thomsonii.
  • Important aroma compound found in Neotinea ustulata.