Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Nonadecene 99.0+%, TCI America™
SDP

Catalog No. S03491ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03491ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03491ML Supplier TCI America Supplier No. S03491ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18435-45-5
Molecular Formula C19H38
Molecular Weight (g/mol) 266.51
MDL Number MFCD00009010
InChI Key NHLUYCJZUXOUBX-UHFFFAOYSA-N
PubChem CID 29075
ChEBI CHEBI:64503
IUPAC Name nonadec-1-ene
SMILES CCCCCCCCCCCCCCCCCC=C

Specifications

Physical Form Crystal/Lump
Color White
Quantity 1 mL
Formula Weight 266.51
Percent Purity ≥99.0% (GC)
Chemical Name or Material 1-Nonadecene [Standard Material for GC]
EINECSNumber (2)-0027
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.