Learn More
Medchemexpress LLC 1-Nonanol | 143-08-8 | 99.1% | 144.26 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYW008816
Encompass_Preferred
Nonan-1-ol is an organic compound used in the manufacture of surfactants and lubricants. It can be used as a detergent, emulsifier, and stabilizer, and is widely applied in cosmetics, personal care products, and printing inks. It also has industrial applications, such as in the production of plastics and rubber.
  • Suitable as a biological material or organic compound
  • Applicable for life science-related research
Catalog No. 50-004-51330