Learn More
Medchemexpress LLC 1-O-1'-(Z)-octadecenyl-2-hydroxy-sn-glycero-3-phosphocholine | 97802-55-6 | 99.0% | 507.3689 Da | C26H54NO6P | 5 MG
SDP
Catalog No. 5000300484
Encompass_Preferred
C18(Plasm) LPC (1-O-1'-(Z)-octadecenyl-2-hydroxy-sn-glycero-3-phosphocholine) is a lysophosphatidylcholine-type plasmalogen used as an endogenous metabolite standard in lipidomics and biochemical research. It is supplied in small research pack sizes and is reported with high purity for analytical applications; distributor listings indicate storage under frozen conditions.
  • High purity (≥99.0%) for analytical reliability.
  • Suitable as an endogenous metabolite standard in lipidomics.
  • Provided in small research pack sizes for analytical workflows.
  • Stored refrigerated or frozen to preserve sample stability.
  • Applicable for method development and reference comparisons.
Catalog No. 50-003-00484 Supplier Medchemexpress LLC Supplier No. HYW0208095MG
C18(Plasm) LPC (1-O-1'-(Z)-octadecenyl-2-hydroxy-sn-glycero-3-phosphocholine) is a lysophosphatidylcholine-type plasmalogen used as an endogenous metabolite standard in lipidomics and biochemical research. It is supplied in small research pack sizes and is reported with high purity for analytical applications; distributor listings indicate storage under frozen conditions.
  • High purity (≥99.0%) for analytical reliability.
  • Suitable as an endogenous metabolite standard in lipidomics.
  • Provided in small research pack sizes for analytical workflows.
  • Stored refrigerated or frozen to preserve sample stability.
  • Applicable for method development and reference comparisons.