Learn More
Cayman Chemical 1-O-Acetylbritannilactone, Purity: ≥98%, CAS: 33627-41-7, 5 mg
SDP
Catalog No. 502910470
Encompass_Preferred
A sesquiterpene lactone with diverse biological activities; inhibits LPS-induced production of PGE2 and nuclear translocation of NF-κB in primary rat aorta VSMCs; inhibits VEGF-induced capillary tube formation in HUVECs at 5 and 10 µM; reduces tumor growth in an A549 mouse xenograft model at 12 mg/kg per day
Catalog No. 50-291-0470 Supplier Cayman Chemical Supplier No. 351375 MG
A sesquiterpene lactone with diverse biological activities; inhibits LPS-induced production of PGE2 and nuclear translocation of NF-κB in primary rat aorta VSMCs; inhibits VEGF-induced capillary tube formation in HUVECs at 5 and 10 µM; reduces tumor growth in an A549 mouse xenograft model at 12 mg/kg per day