Learn More
Cayman Chemical 1-O-Dodecyl-rac-glycerol, Purity: ≥95%, CAS: 1561-07-5, 500 mg
SDP
Catalog No. 502965384
Encompass_Preferred
An alkyl glyceryl ether; a nonionic surfactant; increases autolysin activity in S. faecium and inhibits peptidoglycan synthesis in S. mutans; induces mouse peritoneal macrophage activation ex vivo at 100 ng/animal; increases survival in an Ehrlich murine ascites tumor model at 4 and 8 mg/animal
Catalog No. 50-296-5384 Supplier Cayman Chemical Supplier No. 17441500 MG
An alkyl glyceryl ether; a nonionic surfactant; increases autolysin activity in S. faecium and inhibits peptidoglycan synthesis in S. mutans; induces mouse peritoneal macrophage activation ex vivo at 100 ng/animal; increases survival in an Ehrlich murine ascites tumor model at 4 and 8 mg/animal