Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-O-Hexadecyl-sn-glycerol | 506-03-6 | 99.9% | 316.53 | 100 MG
SDP

Supplier:  Medchemexpress LLC HYW011309A100MG

Encompass_Preferred

1-O-Hexadecyl-sn-glycerol [(S)-HXDG] is an alkyl glycerol monoether derived from Tethya aurantia. It is intended for research use only.

  • Alkyl glycerol monoether
  • Derived from Tethya aurantia
  • Purity of 99.94%
  • Molecular weight of 316.53
  • Formula C19H40O3
  • Appears as a white to off-white solid
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-52864


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.